Learn More
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
£182.00 - £944.00
Product Code | Brand | Capacity (Metric) Cell | Capacity (Metric) Media | For Use With (Application) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) Cell | Capacity (Metric) Media | For Use With (Application) | Includes | Price | Quantity & Availability | ||
12871793
|
DWK Life Sciences
WCL1000-3 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | User manual with feeding and harvesting schedules |
£753.00
Pack of 3 |
|
12801803
|
DWK Life Sciences
WCL1000-1 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | User manual with feeding and harvesting schedules |
£241.00
Each |
09-02-2021 |
12891793
|
DWK Life Sciences
WCL1000AD-1 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for adherent cultures | User manual with feeding and harvesting schedules |
£232.00
Each |
16-02-2021 |
12881793
|
DWK Life Sciences
WCL0350-5 |
5 mL | 350 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | User manual with feeding and harvesting schedules |
£944.00
Pack of 5 |
16-02-2021 |
12811803
|
DWK Life Sciences
WCL1000AD-3 |
15 mL | 1000 mL | Membrane cell culture flasks ideal for antibody and Protein production, for adherent cultures | User manual with feeding and harvesting schedules |
£723.00
Pack of 3 |
16-02-2021 |
12821803
|
DWK Life Sciences
WCL0350-1 |
5 mL | 350 mL | Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | User manual with feeding and harvesting schedules |
£182.00
Each |
16-02-2021 |
Description
High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.
- Bulk media chamber allows for longer run times without media refreshing
- Cell compartment concentrates cells, antibodies and proteins for simplified purification
- For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.
Specifications
Sterile |