missing translation for 'onlineSavingsMsg'
Learn More

Famciclovir, MedChemExpress

Product Code. 16407306
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16407306

Brand: Medchem Express HY17426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Famciclovir(BRL 42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections.

Famciclovir(BRL 42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections.

Specifications

Chemical Name or Material Famciclovir
CAS 104227-87-4
Color White
Purity Grade Notes Research
Molecular Formula C14H19N5O4
Quantity 100 mg
Synonym BRL 42810
Solubility Information DMSO : ≥ 100 mg/mL (311.21 mM) ∣H2O : 50 mg/mL (155.60 mM; Need ultrasonic)
SMILES NC1=NC=C2N=CN(CCC(COC(C)=O)COC(C)=O)C2=N1
Molecular Weight (g/mol) 321.33
Formula Weight 321.33
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.