missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Famciclovir, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Famciclovir(BRL 42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections.
Specifications
Specifications
| Chemical Name or Material | Famciclovir |
| CAS | 104227-87-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H19N5O4 |
| Quantity | 50 mg |
| Synonym | BRL 42810 |
| Solubility Information | DMSO : ≥ 100 mg/mL (311.21 mM) ∣H2O : 50 mg/mL (155.60 mM; Need ultrasonic) |
| SMILES | NC1=NC=C2N=CN(CCC(COC(C)=O)COC(C)=O)C2=N1 |
| Molecular Weight (g/mol) | 321.33 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering