missing translation for 'onlineSavingsMsg'
Learn More

Famciclovir, MedChemExpress

Product Code. 16423357
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423357 50 mg 50mg
16407306 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16423357 Supplier Medchem Express Supplier No. HY17426
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Famciclovir(BRL 42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections.

Famciclovir(BRL 42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections.

Specifications

Chemical Name or Material Famciclovir
CAS 104227-87-4
Color White
Purity Grade Notes Research
Molecular Formula C14H19N5O4
Quantity 50 mg
Synonym BRL 42810
Solubility Information DMSO : ≥ 100 mg/mL (311.21 mM) ∣H2O : 50 mg/mL (155.60 mM; Need ultrasonic)
SMILES NC1=NC=C2N=CN(CCC(COC(C)=O)COC(C)=O)C2=N1
Molecular Weight (g/mol) 321.33
Formula Weight 321.33
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.