missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM98B Recombinant Rabbit Monoclonal Antibody (K01_1R05)

Product Code. 30830124
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830124 100 μL
30829571 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830124 Supplier Invitrogen™ Supplier No. MA563820
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: QNKKHKNSQL DKNSEVYQEV QAMFDTLGIP KSTTSDIPHM LNQVESKVKD ILSKVQKNHV GKPLLKMDLN SEQAEQLERI NDALSCEYEC RRRMLMKRLD VTVQSFGWSD RAKVKTDDIA RIYQPKRYAL SPKTT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 49%, Pig - 93%, Cynomolgus monkey - 97%.

FAM98B positively stimulates PRMT1-induced protein arginine dimethylated arginine methylation. Promotes colorectal cancer cell malignancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM98B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1R05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene FAM98B
Gene Accession No. Q52LJ0
Gene Alias 2610510H03Rik; Fam98b; family with sequence similarity 98 member B; family with sequence similarity 98, member B; protein FAM98B; RGD1564603
Gene Symbols FAM98B
Host Species Rabbit
Immunogen Recombinant protein of human FAM98B (aa 131-265).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283742
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.