missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM98A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | FAM98A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM98A Polyclonal specifically detects FAM98A in Human samples. It is validated for Western Blot.Specifications
| FAM98A | |
| Polyclonal | |
| Rabbit | |
| Q8NCA5 | |
| 25940 | |
| Synthetic peptides corresponding to FAM98A(family with sequence similarity 98, member A) The peptide sequence was selected from the middle region of FAM98A. Peptide sequence QKRQDGPQQQTGGRGGGRGGYEHSSYGGRGGHEQGGGRGGRGGYDHGGRG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZP564F0522, DKFZp686O03192, family with sequence similarity 98, member A, hypothetical protein LOC25940 | |
| FAM98A | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title