missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM98A Polyclonal specifically detects FAM98A in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | FAM98A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | DKFZP564F0522, DKFZp686O03192, family with sequence similarity 98, member A, hypothetical protein LOC25940 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human FAM98A. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?