missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM96B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20108-1-AP-20UL
This item is not returnable.
View return policy
Description
Signs of Rubinstein-Taybi include mental retardation and predisposition to tumor growth and white blood cell neoplasias. Crohn′s disease is a gastrointestinal inflammatory condition associated with chromosome 16 through the NOD2 gene. An association with systemic lupus erythematosis and a number of other autoimmune disorders with the pericentromeric region of chromosome 16 has led to the identification of SLC5A11 as a potential autoimmune modifier. The FAM96B gene product has been provisionally designated FAM96B pending further characterization.Specifications
| FAM96B | |
| Polyclonal | |
| Unconjugated | |
| FAM96B | |
| CGI 128, CIA2B, FAM96B, HSPC118, MIP18, Protein FAM96B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51647, 680987, 68523 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D187, Q9Y3D0 | |
| FAM96B | |
| FAM96B Fusion Protein Ag13827 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction