missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM91A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27738-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM91A1 (family with sequence similarity 91, member A1) is a 94 kDa phospho-protein that is not yet characterized.Specifications
| FAM91A1 | |
| Polyclonal | |
| Unconjugated | |
| FAM91A1 | |
| FAM91A1, Protein FAM91A1, UPF0669 protein C6orf120 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 157769 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q658Y4 | |
| FAM91A1 | |
| FAM91A1 Fusion Protein Ag26902 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction