missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM84B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18421-1-AP-20UL
This item is not returnable.
View return policy
Description
FAM84B is a protein coding gene.Specifications
| FAM84B | |
| Polyclonal | |
| Unconjugated | |
| FAM84B | |
| FAM84B, NSE2, Protein FAM84B, Protein NSE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 157638 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96KN1 | |
| FAM84B | |
| FAM84B Fusion Protein Ag13243 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction