missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM84B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562240
This item is not returnable.
View return policy
Description
Immunogen sequence: EVECSVFYRD ECIYQKSFAP GSAALSTYTP ENLLNKCKPG DLVEFVSQAQ YPHWAVYVGN FQVVHLHRLE VINSFLTDAS Q Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
FAM84B is a protein coding gene.
Specifications
| FAM84B | |
| Polyclonal | |
| Unconjugated | |
| FAM84B | |
| BCMP101; BF642811; breast cancer membrane protein 101; breast cancer membrane-associated protein 101; D330050I23Rik; FAM84B; family with sequence similarity 84 member B; family with sequence similarity 84, member B; LRAT domain-containing 2; LRATD2; neurological/sensory 2; NSE2; Protein FAM84B; Protein LRATD2; Protein NSE2; RGD1562236 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 157638 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96KN1 | |
| FAM84B | |
| Recombinant protein corresponding to Human FAM84B. Recombinant protein control fragment (Product #RP-93893). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction