missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM84B Monoclonal Antibody (OTI1D12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525310
This item is not returnable.
View return policy
Description
FAM84B Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
FAM84B is a protein coding gene.
Specifications
| FAM84B | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96KN1 | |
| FAM84B | |
| Full length human recombinant protein of FAM84B produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1D12 | |
| Unconjugated | |
| FAM84B | |
| BCMP101; BF642811; breast cancer membrane protein 101; breast cancer membrane-associated protein 101; D330050I23Rik; FAM84B; family with sequence similarity 84 member B; family with sequence similarity 84, member B; LRAT domain-containing 2; LRATD2; neurological/sensory 2; NSE2; Protein FAM84B; Protein LRATD2; Protein NSE2; RGD1562236 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 157638 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction