missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM83F Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566703
This item is not returnable.
View return policy
Description
Immunogen sequence: SYRFTWSSSHV DRNLLLLLTG QNVEPFDTEF RELYAISEEV DLYRQLSLAG RVGLHYSSTV ARKLIN Highest antigen sequence identity to the following orthologs - mouse 82%, rat 85%.
FAM83F is a protein coding gene.
Specifications
| FAM83F | |
| Polyclonal | |
| Unconjugated | |
| FAM83F | |
| AW544981; FAM83F; family with sequence similarity 83 member F; family with sequence similarity 83, member F; protein FAM83F; RGD1562089 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEG4 | |
| FAM83F | |
| Recombinant Human FAM83F. Recombinant protein control fragment (Product #RP-107354). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction