missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM83E Polyclonal Antibody

Description
Immunogen sequence: DNELKKSWGS KDTPAKALMR QRGTGGGPWG EVDSRPPWGG ALPLPPAHRL RYLSPARRRF GGDATFKLQE P Antigen sequence identity: Mouse 67%; Rat 65%.
FAM83E may play a role in MAPK signaling.
Specifications
Specifications
| Antigen | FAM83E |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FAM83E |
| Gene Accession No. | Q2M2I3 |
| Gene Alias | 4930403C10Rik; FAM83E; family with sequence similarity 83 member E; family with sequence similarity 83, member E; protein FAM83E; RGD1309316 |
| Gene Symbols | FAM83E |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?