missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM81B Recombinant Mouse Monoclonal Antibody (K40071_18B11)

Product Code. 30829609
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829609 50 μL
30829329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829609 Supplier Invitrogen™ Supplier No. MA565880
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: EMNLLEFKFH SLSSNLYEEV ENNKKWTENQ FLKYRKDHLG HINECLKVLQ EKLEKSENKM EEKLLQLSSK VENFINTQKQ ETQLSKVKHM ENKLSKKMEQ MEKQIWGELE Highest antigen sequence identity to the following orthologs: Pig - 76%, Cynomolgus monkey - 98%.

FAM81B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM81B
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K40071_18B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene FAM81B
Gene Accession No. Q96LP2
Gene Alias FAM81B; family with sequence similarity 81 member B; family with sequence similarity 81, member B; Gm276; Protein FAM81B; RGD1561830
Gene Symbols FAM81B
Host Species Mouse
Immunogen Recombinant human FAM81B (aa 291-400).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153643
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.