missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM76B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117946
This item is not returnable.
View return policy
Description
FAM76B Polyclonal Antibody for Western Blot, IP
FAM76B is an uncharacterized protein belonging to the FAM76 family.
Specifications
| FAM76B | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q5HYJ3 | |
| Fam76b | |
| E. coli-derived Human FAM76B fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Fam76b | |
| 2810485I05Rik; C78303; FAM76B; family with sequence similarity 76 member B; family with sequence similarity 76, member B; protein FAM76B; RGD1311077 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 143684 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction