missing translation for 'onlineSavingsMsg'
Learn More

FAM73B Polyclonal Antibody, Invitrogen™

Product Code. 15737391 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15737391 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15737391 Supplier Invitrogen Supplier No. PA559535

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RTLMLPLTEG SLRLRADDED SLTSEDSFFS ATELFESLQT GDYPIPLSRP AAAYEEALQL VKEGRVPCRT LRTELLGCYS DQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM73B
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIGA2
Gene Accession No. Q7L4E1
Gene Alias C9orf54, protein FAM73B
Gene Symbols MIGA2
Host Species Rabbit
Immunogen Recombit protein corresponding to Human FAM73B
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.