Learn More
FAM71F2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24286-1-AP-150UL
 View more versions of this product
Product Code. 16892755
  • £349.00 / 150µL
Specifications
FAM71F2
Polyclonal
Unconjugated
FAM71F2
FAM137B, FAM71F2, Protein FAM137B, Protein FAM71F2
Rabbit
Antigen Affinity Chromatography
RUO
245884, 346653
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.66 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6NXP2
FAM71F2
FAM71F2 Fusion Protein Ag16874
150 μL
Primary
Human, Mouse