missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM71B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559291
This item is not returnable.
View return policy
Description
Immunogen sequence: QELGKNQSAS STGALQKKAS KISSFLRSLR ATPGSKTRVT SHDREVDIVA KMVEKQNIEA KVEKAQGGQE LEMISGTMTS EKTEMIVFET Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 47%.
FAM71B is a protein coding gene.
Specifications
| FAM71B | |
| Polyclonal | |
| Unconjugated | |
| FAM71B | |
| FAM71B; family with sequence similarity 71 member B; family with sequence similarity 71, member B; HSD51; HSD-51; Protein FAM71B; testicular tissue protein Li 68; Unknown (protein for IMAGE:8061995) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 153745 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TC56 | |
| FAM71B | |
| Recombinant protein corresponding to Human FAM71B. Recombinant protein control fragment (Product #RP-96934). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction