missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM65C Polyclonal specifically detects FAM65C in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | C20orf175 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q96MK2 |
| Gene Alias | C20orf176, Chromosome 20 Open Reading Frame 175, Chromosome 20 Open Reading Frame 176, dJ530I15.2, dJ530I15.3, FAM65C, Family With Sequence Similarity 65, Member C, Protein FAM65C |
| Gene Symbols | FAM65C |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?