missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM65B, Mouse anti-Human, Clone: 2E6-1B10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009750-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MLVGSQSFSPGGPNGIIRSQSFAGFSGLQERRSRCNSFIENSSALKKPQAKLKKMHNLGHKNNNPPKEPQPKRVEEVYRALKNGLDEYLEVHQTELDKLTAQLKDMKRNSRLGVLYDLDKQIKTIERYMRRLEFHISKVDELYEAYCIQRRLQDGASKMKQAFATSPASKAARESLTEINRSFKEYTENMCTIEVELENLLGEFSIKMKGLAGFARLCPGDQYEIFMKYGRQRWKLKGKIEVNGKQSWDGEETVFLPLIVGFISIKVTELKGLATHILVGSVTCETKELFAARPQVVAVDINDLGTIKLNLEITWYPFDVEDMTASSGAGNKAAALQRRMSMYSQGTPETPTFKDHSFFSNLPDDIFENGKAAEEKMPLSLSFSDLPNGDCALTSHSTGSPSNSTNPEITITPAEFNLSSLASQNEGMDDTSSASSRNSLGEGQEPKSHLKEEDPEEPRKPASAPSEACRRQSSGAGAEHLFLENDVAEALLQESEEASELKPVELDTSEGNITKQLVKRLTSAEVPMATDRLLSEGSVGGESEGCRSFLDGSLEDAFNGLLLALEPHKEQYKEFQDLNQEVMNLDDILKKSpecifications
| FAM65B | |
| Monoclonal | |
| Unconjugated | |
| family with sequence similarity 65, member B | |
| C6orf32/DIFF40/DIFF48/KIAA0386/PL48 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2E6-1B10 | |
| In 1x PBS, pH 7.4 | |
| BC001232 | |
| FAM65B | |
| C6orf32 (AAH01232.1, 1 a.a. ∼ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9750 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction