missing translation for 'onlineSavingsMsg'
Learn More

FAM65B Antibody (2F6-1A11), Novus Biologicals™

Product Code. 18367198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367198 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367198 Supplier Novus Biologicals Supplier No. H00009750M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FAM65B Monoclonal antibody specifically detects FAM65B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM65B
Applications Western Blot, Flow Cytometry, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2F6-1A11
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry, ELISA 1:100-1:2000, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01232
Gene Alias C6orf32, DIFF48DIFF40, family with sequence similarity 65, member B, PL48KIAA0386chromosome 6 open reading frame 32
Host Species Mouse
Immunogen C6orf32 (AAH01232.1, 1 a.a. ~ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLVGSQSFSPGGPNGIIRSQSFAGFSGLQERRSRCNSFIENSSALKKPQAKLKKMHNLGHKNNNPPKEPQPKRVEEVYRALKNGLDEYLEVHQTELDKLTAQLKDMKRNSRLGVLYDLDKQIKTIERYMRRLEFHISKVDELYEAYCIQRRLQDGASKMKQAFATSPASKAARESLTEINRSFKEYTENMCTIEVELENLLGEFSIKMKGLAGFARLCPGDQYEIFMKYGRQRWKLKGKIEVNGKQSWDGEETVFLPLIVGFISIKVTELKGLATHILVGSVTCETKELFAARPQVVAVDINDLGTIKLNLEITWYPFDVEDMTASSGAGNKAAALQRRMSMYSQGTPETPTFKDHSFFSNLPDDIFENGKAAEEKMPLSLSFSDLPNGDCALTSHSTGSPSNSTNPEITITPAEFNLSSLASQNEGMDDTSSASSRNSLGEGQEPKSHLKEEDPEEPRKPASAPSEACRRQSSGAGAEHLFLENDVAEALLQESEEASELKPVELDTSEGNITKQLVKRLTSAEVPMATDRLLSEGSVGGESEGCRSFLDGSLEDAFNGLLLALEPHKEQYKEFQDLNQEVMNLDDILKK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9750
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.