missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM65A Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78755AF488
This item is not returnable.
View return policy
Description
FAM65A Polyclonal antibody specifically detects FAM65A in Human samples. It is validated for Western Blot
Specifications
| FAM65A | |
| Polyclonal | |
| Western Blot | |
| family with sequence similarity 65, member A, hypothetical protein LOC79567 | |
| The immunogen for this product maps to a region between residue 1173 and 1223 of human Family with Sequence Similarity 65, Member A using the numbering given in entry NP_001180451.1 (GeneID 79567). | |
| 0.1 mL | |
| Cell Biology | |
| 79567 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction