missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM62B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109710
This item is not returnable.
View return policy
Description
Immunogen sequence: EKRERPPDHQ HSAQVKRPSV SKEGRKTSIK SHMSGSPGPG GSNTAPSTPV IGGSDKPGME EKAQPPEAGP QGLHDLGRSS SSLLASPGHI SVKEPTPSIA SDISLPIATQ ELRQRLRQLE NGTTLGQSPL G.
Tethers the endoplasmic reticulum to the cell membrane and promotes the formation of appositions between the endoplasmic reticulum and the cell membrane. Binds glycerophospholipids in a barrel-like domain and may play a role in cellular lipid transport. Plays a role in FGF signaling via its role in the rapid internalization of FGFR1 that has been activated by FGF1 binding; this occurs most likely via the AP-2 complex. Promotes the localization of SACM1L at endoplasmic reticulum-plasma membrane contact sites (EPCS) (PubMed:27044890). [UniProt].
Specifications
| FAM62B | |
| Polyclonal | |
| Unconjugated | |
| ESYT2 | |
| 2410017M09Rik; 4921504I16Rik; chr2 synaptotagmin; CHR2SYT; D12Ertd551e; ESYT2; E-Syt2; extended synaptotagmin 2; extended synaptotagmin like protein 2; extended synaptotagmin protein 2; extended synaptotagmin-2; extended synaptotagmin-like protein 2; Fam62b; family with sequence similarity 62 (C2 domain containing), member B; family with sequence similarity 62, member B; KIAA1228; RGD1565705 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 52635, 57488 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| A0FGR8, Q3TZZ7 | |
| ESYT2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 630-760 of human FAM62B (NP_065779.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction