missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM5C Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £366.00
Specifications
| Antigen | FAM5C |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677060
|
Novus Biologicals
NBP2-93578-0.02ml |
0.02 mL |
£186.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623750
|
Novus Biologicals
NBP2-93578-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM5C Polyclonal antibody specifically detects FAM5C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FAM5C | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 339479 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DBCCR1LDBCCR1L1BRINP3, DBCCR1-like protein 1, family with sequence similarity 5, member C, RP11-445K1.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 240-500 of human FAM5C (NP_950252.1). LQGLQVLLPDYLQERFVQAALSYIACNSEGEFICKENDCWCHCGPKFPECNCPSMDIQAMEENLLRITETWKAYNSDFEESDEFKLFMKRLPMNYFLNTSTIMHLWTMDSNFQRRYEQLENSMKQLFLKAQKIVHKLFSLSKRCHKQPLISLPRQRTSTYWLTRIQSFLYCNENGLLGSFSEETHSCTCPNDQVVCTAFLPCTVGDASACLTCAPDNRTRCGTCNTGYMLSQGLCKPEVAESTDHYIGFETDLQDLEMKYL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title