missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM58A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562142
This item is not returnable.
View return policy
Description
Immunogen sequence: GVEVPAEVEA EKPWWQVFND DLTKPIIDNI VSDLIQIYTM DTE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Activating cyclin for the cyclin-associated kinase CDK10.
Specifications
| FAM58A | |
| Polyclonal | |
| Unconjugated | |
| CCNQ | |
| Ccnq; CDK10-activating cyclin; cyclin M; cyclin Q; cyclin-M; Cyclin-Q; Cyclin-related protein FAM58A; Fam58a; Fam58b; family with sequence similarity 58 member A; family with sequence similarity 58, member A; family with sequence similarity 58, member B; RGD1305651; STAR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92002 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1B3 | |
| CCNQ | |
| Recombinant protein corresponding to Human FAM58A. Recombinant protein control fragment (Product #RP-98487). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction