missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM54A Monoclonal Antibody (OTI2G8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527461
This item is not returnable.
View return policy
Description
FAM54A Monoclonal Antibody for Western Blot
FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.
Specifications
| FAM54A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B0K035, Q6P444 | |
| MTFR2 | |
| Full length human recombinant protein of FAM54A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| MTFR2 | |
| DUF729 domain containing 1; DUF729 domain-containing protein 1; DUFD1; EGK_15321; FAM54A; family with sequence similarity 54, member A; LOC100911069; Mitochondrial fission regulator 2; Mtfr2; protein FAM54A; protein FAM54A-like | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100911069, 113115, 483997 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction