missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM53B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112903
This item is not returnable.
View return policy
Description
FAM53B Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| FAM53B | |
| Polyclonal | |
| Unconjugated | |
| FAM53B | |
| A930008G19Rik; bA12J10.2; FAM53B; family with sequence similarity 53 member B; family with sequence similarity 53, member B; KIAA0140; mKIAA0140; Protein FAM53B; Protein simplet; RGD1310799; RP11-12J10.2; simplet homolog; smp | |
| Rabbit | |
| Protein G | |
| RUO | |
| 9679 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q14153 | |
| FAM53B | |
| Recombinant Human Protein FAM53B protein (1-305AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction