missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM53A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557666
This item is not returnable.
View return policy
Description
Immunogen sequence: LDDLTCKAEA GPLQYSAETL NKSGRLFPLE LNDQSPWKVF SGGPPVRSQA ATGPDFSFLP GLSAAAHTMG LQWQPQSPRP GAGLGAASTV DPSEST Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 52%.
Dorsal neural-tube nuclear protein is a 398 amino acid nuclear protein. DNTNP is thought to play an important role in neural development by specifying dorsal cell fates within the neural tube. Widely expressed in the dorsal neural tube, DNTNP is most highly expressed in the dorsal regions of the midbrain, the hindbrain, the diencephalon, and the spinal neural tube and is expressed at lower levels in the branchial arches, the telencephalon, the heart, and somites of developing embryos. DNTNP is encoded by a gene located on chromosome 4 which contains many genes including the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease.
Specifications
| FAM53A | |
| Polyclonal | |
| Unconjugated | |
| FAM53A | |
| DNTNP; Dorsal neural-tube nuclear protein; FAM53A; family with sequence similarity 53 member A; family with sequence similarity 53, member A; protein FAM53A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 152877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NSI3 | |
| FAM53A | |
| Recombinant protein corresponding to Human FAM53A. Recombinant protein control fragment (Product #RP-96742). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction