missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM53A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | FAM53A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FAM53A Polyclonal specifically detects FAM53A in Human samples. It is validated for Western Blot.Specifications
| FAM53A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DNTNP, Dorsal neural-tube nuclear protein, family with sequence similarity 53, member A | |
| FAM53A | |
| IgG | |
| 42 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q6NSI3 | |
| 152877 | |
| Synthetic peptides corresponding to FAM53A (family with sequence similarity 53, member A) The peptide sequence was selected from the C terminal of FAM53A. Peptide sequence RGLRTSPVHPNLWASRESVTSDGSRRSSGDPRDGDSVGEEGVFPRARWEL. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title