missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WDR5 Recombinant Human Monoclonal Antibody (RAB-C223)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551835
This item is not returnable.
View return policy
Description
This antibody recognizes WDR5 (WD repeat-containing protein 5). It binds to a folded domain, amino acids 1-334. WDR5 is a protein that is involved in histone modification.
Cytosolic prostaglandin synthase that catalyzes the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Facilitates HIF alpha proteins hydroxylation via interaction with EGLN1/PHD2, leading to recruit EGLN1/PHD2 to the HSP90 pathway.
Specifications
| WDR5 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P61964 | |
| WDR5 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing WDR5 protein under non-denaturing conditions. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| F(ab) κ |
| ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Mass Spectrometry | |
| RAB-C223 | |
| Unconjugated | |
| WDR5 | |
| 2410008O07Rik; AA408785; AA960360; Big; BIG3; BIG-3; BMP2-induced 3-kb gene protein; CFAP89; cilia and flagella associated protein 89; SWD3; SWD3, Set1c WD40 repeat protein, homolog; Unknown (protein for MGC:137327); WD repeat domain 5; WD repeat-containing protein 5; WD repeat-containing protein BIG-3; Wdr5; wu:fk47f04; zgc:56591; zgc:76895; zgc:76895 protein | |
| Human | |
| Metal-chelate chromatography | |
| RUO | |
| 11091 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction