missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM49A Polyclonal Antibody
FAM49A Polyclonal Antibody
Brand: Invitrogen PA5107121
This item is not returnable.
View return policy
Description
FAM49A Polyclonal Antibody for Western Blot
Specifications
| FAM49A | |
| Polyclonal | |
| Unconjugated | |
| FAM49A | |
| 2410157M17Rik; 9630047E15; D12Ertd553e; FAM49A; family with sequence similarity 49 member A; family with sequence similarity 49, member A; family with sequence similarity 49, member B; protein FAM49A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 81553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9H0Q0 | |
| FAM49A | |
| Synthetic peptide directed towards the C-terminal region of Human FAM49A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction