missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: RSPCNLAIPS EVDVEKYAKV EKSIKSDDSQ PTVWPAHDVK DDYVFECEAG TQYQKTKLTI LQSLGDPLYY GKIQPCKADE ESDSQMSPSH SSTDD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
Required for MAP kinase p38 (MAPK11, MAPK12, MAPK13 and/or MAPK14) activation during gastrulation. Required for down-regulation of E-cadherin during gastrulation by regulating E-cadherin protein level downstream from NCK-interacting kinase (NIK) and independently of the regulation of transcription by FGF signaling and Snail. Required for starvation-induced ATG9A trafficking during autophagy. [UniProt].
Specifications
Specifications
| Antigen | FAM48A |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SUPT20H |
| Gene Accession No. | Q8NEM7 |
| Gene Alias | bA421P11.4; C13; C13orf19; Fam48a; family with sequence similarity 48, member A; FP757; p38-interacting protein; P38IP; protein FAM48A; RGD1307812; SPT20; SPT20 homolog, SAGA complex component; suppressor of Ty 20; suppressor of Ty 20 homolog; Supt20; SUPT20H; transcription factor (p38 interacting protein); transcription factor SPT20 homolog; tumor rejection antigen |
| Gene Symbols | SUPT20H |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
