missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM46C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544108
This item is not returnable.
View return policy
Description
Peptide sequence: LIATKNPEEI RGGGLLKYSN LLVRDFRPTD QEEIKTLERY MCSRFFIDFP Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
FAM46C belongs to the FAM46 family. The exact function of FAM46C remains unknown.
Specifications
| FAM46C | |
| Polyclonal | |
| Unconjugated | |
| FAM46C | |
| 4930431B09Rik; AI449797; DN-384L23.3; Fam46c; family with sequence similarity 46 member C; family with sequence similarity 46, member C; Non-canonical poly(A) polymerase FAM46C; protein FAM46C; Protein FAM46C-like protein; putative nucleotidyltransferase FAM46C; TENT5C; Terminal nucleotidyltransferase 5C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54855 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5VWP2 | |
| FAM46C | |
| Synthetic peptide directed towards the middle region of human FAM46C. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction