missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM46A Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99201CL1
This item is not returnable.
View return policy
Description
FAM46A Polyclonal antibody specifically detects FAM46A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FAM46A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| C6orf37, chromosome 6 open reading frame 37, family with sequence similarity 46, member A, FLJ20037, FLJ31495, HBV XAg-transactivated protein 11, HBV X-transactivated gene 11 protein, hypothetical protein LOC55603, retinal expressed gene C6orf37, XTP11 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FAM46A. | |
| 0.1 mL | |
| Cell Biology | |
| 55603 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction