missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM44A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559504
This item is not returnable.
View return policy
Description
Immunogen sequence: KSEDTQKVKD EKQAKEKEVE SLKLPSEKNS NKAKTVEGTK EDFSLIDSDV DGLTDITVSS VHTSDLSSFE EDTEEEVVT Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
The FAM44A gene, also known as Family With Sequence Similarity 44 Member A, is located on chromosome 10q11.22. This gene encodes a protein that is implicated in cellular processes but its precise biological function remains to be fully elucidated. FAM44A's expression is observed predominantly in the testes, and it is believed to potentially play a role in spermatogenesis. Structurally, it includes various exons and introns characteristic of protein-coding genes. FAM44A is considered part of poorly characterized genes, and ongoing research is aimed at understanding its functional dynamics, potential roles in disease conditions, and interactions with other proteins. Databases like STRING suggest protein-protein interactions involving FAM44A, indicating its participation in complex cellular networks.
Specifications
| FAM44A | |
| Polyclonal | |
| Unconjugated | |
| BOD1L1 | |
| biorientation of chromosomes in cell division 1 like 1; biorientation of chromosomes in cell division 1-like 1; biorientation of chromosomes in cell division protein 1-like 1; BOD1L; BOD1L1; FAM44A; family with sequence similarity 44, member A; KIAA1327 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 259282 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFC6 | |
| BOD1L1 | |
| Recombinant protein corresponding to Human FAM44A. Recombinant protein control fragment (Product #RP-96593). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction