missing translation for 'onlineSavingsMsg'
Learn More

FAM44A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA557918

Additional Details : Weight : 0.02000kg

Product Code. 15725811

  • £427.00 / 100µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: PQVEKAVHEF LATLNHKEEG SGNTAPDDEK PDTSLITQGV PTPGPSANVA NDAMSILETI TSLNQEASAA RASTETSNAK TSE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

FAM44A Polyclonal antibody specifically detects FAM44A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

FAM44A
Polyclonal
Unconjugated
BOD1L1
biorientation of chromosomes in cell division 1 like 1; biorientation of chromosomes in cell division 1-like 1; biorientation of chromosomes in cell division protein 1-like 1; BOD1L; BOD1L1; FAM44A; family with sequence similarity 44, member A; KIAA1327
Rabbit
Antigen affinity chromatography
RUO
259282
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NFC6
BOD1L1
Recombinant protein corresponding to Human FAM44A
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only