missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM43A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113287
This item is not returnable.
View return policy
Description
FAM43A Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| FAM43A | |
| Polyclonal | |
| Unconjugated | |
| FAM43A | |
| Fam43a; family with sequence similarity 43 member A; family with sequence similarity 43, member A; PP7298; Protein FAM43A; somitovasculin; Tuf1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 131583, 224093 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BUP8, Q8N2R8 | |
| FAM43A | |
| Recombinant Human Protein FAM43A protein (1-200AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction