missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM40A Monoclonal Antibody (OTI7G7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525394
This item is not returnable.
View return policy
Description
FAM40A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the cortical actin filament dynamics and cell shape.
Specifications
| FAM40A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5VSL9 | |
| STRIP1 | |
| Full length human recombinant protein of FAM40A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI7G7 | |
| Unconjugated | |
| STRIP1 | |
| 6330569M22Rik; 6530401O14Rik; Fam40a; family with sequence similarity 40, member A; FAR11 factor arrest 11 homolog A; FAR11A; homolog of yeast FAR11 protein 1; hypothetical protein; Kiaa1761; Protein FAM40A; QnpA-18494; RP4-773N10.1; striatin interacting protein 1; striatin-interacting protein 1; striatin-interacting protein 1; LOW QUALITY PROTEIN: striatin-interacting protein 1; Strip1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 362012, 490117, 85369 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction