missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM3D Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5109709

Product Code. 17117643

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RSYMSFSMKT IRLPRWLAAS PTKEIQVKKY KCGLIKPCPA NYFAFKICSG AANVVGPTMC FEDRMIMSPV KNNVGRGLNI ALVNGTTGAV LGQKAFDMYS GDVMHLVKFL KEIPGGALVL VASYDDPGTK MNDESRKLFS DLGSSYAKQL GFRDSWVFIG AKDLRGKSPF EQFLKNSPDT N.

FAM3D (FAM3 Metabolism Regulating Signaling Molecule D) is a Protein Coding gene. Diseases associated with FAM3D include Deafness, Autosomal Recessive 17 and Scheuermann Disease. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is FAM3C. [GeneCards].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM3D
Polyclonal
Unconjugated
Fam3d
2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; Fam3d; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; oncoprotein-induced protein 1; Protein EF-7; protein FAM3D; UNQ567/PRO1130
Rabbit
Affinity Chromatography
RUO
131177, 18300, 289949
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.11 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P97805, Q96BQ1
Fam3d, Oit1
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.