missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM3D Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547434
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) FAM3D is observed and less than 1% cross reactivity with rhFAM3A, rhFAM3B and rhFAM3C is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
FAM3D (FAM3 Metabolism Regulating Signaling Molecule D) is a Protein Coding gene. Diseases associated with FAM3D include Deafness, Autosomal Recessive 17 and Scheuermann Disease. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is FAM3C. [GeneCards].
Specifications
| FAM3D | |
| Polyclonal | |
| Unconjugated | |
| Oit1 | |
| 2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; Fam3d; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; oncoprotein-induced protein 1; Protein EF-7; protein FAM3D; UNQ567/PRO1130 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 18300 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P97805 | |
| Oit1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse FAM3D Trp39-Met224. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction