missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM3D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537466
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 25 kDa. Purity is >95% by SDS-PAGE.
FAM3D (FAM3 Metabolism Regulating Signaling Molecule D) is a Protein Coding gene. Diseases associated with FAM3D include Deafness, Autosomal Recessive 17 and Scheuermann Disease. Gene Ontology (GO) annotations related to this gene include cytokine activity. An important paralog of this gene is FAM3C. [GeneCards].
Specifications
| FAM3D | |
| Polyclonal | |
| Unconjugated | |
| Fam3d | |
| 2310076N21Rik; AV067083; cytokine-like protein EF-7; EF7; EF-7; Fam3d; family with sequence similarity 3 member D; family with sequence similarity 3, member D; Oit1; oncoprotein induced transcript 1; oncoprotein induced transcript 1 homolog; oncoprotein-induced protein 1; Protein EF-7; protein FAM3D; UNQ567/PRO1130 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 131177 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| Q96BQ1 | |
| Fam3d | |
| Synthetic peptide corresponding to amino acids 168-220 of human FAM3D. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction