Learn More
FAM3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14247-1-AP-150UL
 View more versions of this product
Product Code. 16842804
  • £349.00 / 150µL
Description
FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.
Specifications
Specifications
FAM3C
Polyclonal
Unconjugated
FAM3C
FAM3C, GS3786, ILEI, Interleukin like EMT inducer, Protein FAM3C
Rabbit
Antigen Affinity Chromatography
RUO
10447, 27999, 312159
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q810F4, Q91VU0, Q92520
FAM3C
FAM3C Fusion Protein Ag5516
150 μL
Primary
Human, Mouse, Rat