Learn More
FAM3C Mouse anti-Human, CoraLite 488, Clone: 2E10D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60282-100UL
Description
The FAM3C antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FAM3C. This antibody recognizes human antigen. The FAM3C antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| FAM3C | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 10447 | |
| IgG1 |
| Immunofluorescence | |
| 2E10D5 | |
| Mouse | |
| Primary | |
| Human |