missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM3B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27131-1-AP-150UL
This item is not returnable.
View return policy
Description
FAM3B (family with sequence similarity 3, member B), also known as ORF9, C21orf11 or PANDER, is a 235 amino acid secreted protein that localizes to discrete vesicular and perinuclear structures. Expressed at high levels in pancreas and at lower levels in kidney, colon, testis, prostate and small intestine, FAM3B functions as an islet-specific cytokine that promotes apoptosis and may inhibit Insulin secretion from beta-cells, possibly playing a role in overall pancreatic activity. Human FAM3B shares 78% sequence identity with its mouse counterpart, suggesting a conserved role between species. Three isoforms of FAM3B, designated A, B and C, exist due to alternative splicing events.Specifications
| FAM3B | |
| Polyclonal | |
| Unconjugated | |
| FAM3B | |
| C21orf11, C21orf76, Cytokine like protein 2 21, FAM3B, ORF9, Pancreatic derived factor, PANDER, PRED44, Protein FAM3B, UNQ320/PRO365 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54097 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P58499 | |
| FAM3B | |
| FAM3B Fusion Protein Ag25955 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction