missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM3B, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00054097-B02P.50ug
This item is not returnable.
View return policy
Description
Sequence: MRPLAGGLLKVVFVVFASLCAWYSGYLLAELIPDAPLSSAAYSIRSIGERPVLKAPVPKRQKCDHWTPCPSDTYAYRLLSGGGRSKYAKICFEDNLLMGEQLGNVARGINIAIVNYVTGNVTATRCFDMYEGDNSGPMTKFIQSAAPKSLLFMVTYDDGSTRLNNDAKNAIEALGSKEIRNMKFRSSWVFIAAKGLELPSEIQREKINHSDAKNNRYSGWPAEIQIEGCIPKERSSpecifications
| FAM3B | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_058186 | |
| FAM3B | |
| FAM3B (NP_478066.3, 1 a.a. ∼ 235 a.a) full-length human protein. | |
| RUO | |
| 54097 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| family with sequence similarity 3, member B | |
| 2-21/C21orf11/C21orf76/ORF9/PANDER/PRED44 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction