missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | FAM3B |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274492
|
Novus Biologicals
NBP2-56789 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624566
|
Novus Biologicals
NBP2-56789-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAM3B Polyclonal specifically detects FAM3B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| FAM3B | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 54097 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AELIPDAPLSSAAYSIRSIGERPVLKAPV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PRED44 | |
| FAM3B | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title