Learn More
Invitrogen™ FAM222B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114271
Description
Immunogen sequence: VNGMAAPLAY PNGHYFQPLW NNILPTPNSD SSGSQDLAMP FHGGQPTGAP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
FAM222B is a protein coding gene.
Specifications
| FAM222B | |
| Polyclonal | |
| Unconjugated | |
| FAM222B | |
| 9630020I17Rik; C17orf63; FAM222B; family with sequence similarity 222 member B; family with sequence similarity 222, member B; protein FAM222B; uncharacterized protein C17orf63; uncharacterized protein C17orf63 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55731 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WU58 | |
| FAM222B | |
| Synthetic peptide directed towards the C-terminal region of Human FAM222B (aa 435-484). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.