missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAM219B Polyclonal specifically detects FAM219B in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | FAM219B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | C15orf17, chromosome 15 open reading frame 17, FLJ00005, hypothetical protein LOC57184 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human C15ORF17 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?