missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM213B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552290

Product Code. 15750241

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LAGLLDQHGV RLVGVGPEAL GLQEFLDGDY FAGELYLDES KQLYKELGFK RYNSLSILPA ALGKPVRDVA AKAKAVGIQG NLSGDLLQSG GLLVVSKGGD KVLLHFVQKS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

FAM213B is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM213B
Polyclonal
Unconjugated
FAM213B
2810405K02Rik; AI836168; C16H1orf93; C1orf93; C5H1orf93; Fam213b; family with sequence similarity 213 member B; family with sequence similarity 213, member B; hypothetical protein MGC26818; LOW QUALITY PROTEIN: prostamide/prostaglandin F synthase; peroxiredoxin like 2B; Peroxiredoxin-like 2B; PM/PGFS; Prostamide/PG F synthase; prostamide/PGF synthase; prostamide/prostaglandin F synthase; Protein FAM213B; prxl2; Prxl2b; RGD1308251; sb:cb782; selu3; wu:fc33a02; zgc:85644
Rabbit
Antigen affinity chromatography
RUO
127281, 66469
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TBF2, Q9DB60
FAM213B
Recombinant protein corresponding to Human FAM213B. Recombinant protein control fragment (Product #RP-88847).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.